N-Benzyloxycarbonyl-D-alaninol

Proveedor: Apollo Scientific

Sinónimos: (2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol

OR316021-25G
APOSOR316021-25GEA 72.5 EUR
APOSOR316021-25G
N-Benzyloxycarbonyl-D-alaninol
N-Benzyloxycarbonyl-D-alaninol
Formula: C₁₁H₁₅NO₃
MW: 209,2 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00672531
Núm. CAS: 61425-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR