2-(4-Benzylpiperazin-1-yl)benzaldehyde dihydrochloride 95+%

Proveedor: Apollo Scientific

OR318091-1G OR318091-10G OR318091-5G
APOSOR318091-1GEA 208 EUR
APOSOR318091-1G APOSOR318091-10G APOSOR318091-5G
2-(4-Benzylpiperazin-1-yl)benzaldehyde dihydrochloride 95+%
2-(4-Benzylpiperazin-1-yl)benzaldehyde dihydrochloride
Formula: C₁₈H₂₂Cl₂N₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11226767
Núm. CAS: 1170029-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR