sec-Butylhydrazine dihydrochloride 95+%

Proveedor: Apollo Scientific
Peligro

OR318093-1G OR318093-25G OR318093-5G
APOSOR318093-1GEA 166 EUR
APOSOR318093-1G APOSOR318093-25G APOSOR318093-5G
sec-Butylhydrazine dihydrochloride 95+%
sec-Butylhydrazine dihydrochloride
Formula: C₄H₁₄Cl₂N₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11840328
Núm. CAS: 1177361-36-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR