1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one 95%
Proveedor: Apollo Scientific
OR32346-1G
OR32346-5G
APOSOR32346-1GEA
180
EUR
APOSOR32346-1G
APOSOR32346-5G
1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one 95%
1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one
Formula:
C₁₄H₁₇NO₂ MW: 231,3 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08056608 Núm. CAS: 886361-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...