1-(2,6-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
Proveedor: Apollo Scientific
Sinónimos:
1-[(2,6-Dichlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid
OR32955-5G
APOSOR32955-5GEA
366
EUR
APOSOR32955-5G
1-(2,6-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-(2,6-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formula:
C₁₃H₉Cl₂NO₃ MW: 298,13 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00139897 Núm. CAS: 338783-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...