3-(Dimethylamino)-1-phenyl-2-buten-1-one 95%
Proveedor: Apollo Scientific
OR33117-1G
OR33117-5G
APOSOR33117-1GEA
270
EUR
APOSOR33117-1G
APOSOR33117-5G
3-(Dimethylamino)-1-phenyl-2-buten-1-one 95%
3-(Dimethylamino)-1-phenyl-2-buten-1-one
Formula:
C₁₂H₁₅NO MW: 189,26 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08056660 Núm. CAS: 34523-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...