6-Bromo-1,3-benzodioxole-5-carbonitrile 95%
Proveedor: Apollo Scientific
OR33276-5G
APOSOR33276-5GEA
180
EUR
APOSOR33276-5G
6-Bromo-1,3-benzodioxole-5-carbonitrile 95%
6-Bromo-1,3-benzodioxole-5-carbonitrile
Formula:
C₈H₄BrNO₂ MW: 226,04 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02571372 Núm. CAS: 6120-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...