3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]hydrochloride 95%
Proveedor: Apollo Scientific
OR33502-1G
OR33502-5G
APOSOR33502-1GEA
270
EUR
APOSOR33502-1G
APOSOR33502-5G
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]hydrochloride 95%
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]hydrochloride
Formula:
C₁₂H₁₆ClNS MW: 241,79 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD14584833 Núm. CAS: 1221792-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...