3-(Hydroxymethyl)benzoic acid 98%
Proveedor: Apollo Scientific
OR345068-5G
APOSOR345068-5GEA
88.6
EUR
APOSOR345068-5G
3-(Hydroxymethyl)benzoic acid 98%
3-(Hydroxymethyl)benzoic acid
Formula:
C₈H₈O₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06208242 Núm. CAS: 28286-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...