N'-[2-(4-Chlorophenyl)-1-cyclopropylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester
Proveedor: Apollo Scientific
OR346037-1G
OR346037-500MG
APOSOR346037-1GEA
0
EUR
APOSOR346037-1G
APOSOR346037-500MG
N'-[2-(4-Chlorophenyl)-1-cyclopropylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[2-(4-Chlorophenyl)-1-cyclopropylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula:
C₁₆H₂₂ClN₃O₂ MW: 323,83 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD10568250 Núm. CAS: 1053657-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...