N,N'-Dicarbobenzoxy-L-cystine 97%

Proveedor: Apollo Scientific

Sinónimos: (Z-Cystine-OH)₂, (Z-Cys-OH)2, N,N'-Di-Cbz-L-cystine, N,N'-Dibenzyloxycarbonyl-L-cystine

OR452094-25G OR452094-100G
APOSOR452094-25GEA 42.2 EUR
APOSOR452094-25G APOSOR452094-100G
N,N'-Dicarbobenzoxy-L-cystine 97%
N,N'-Dicarbobenzoxy-L-cystine
Formula: C₂₂H₂₄N₂O₈S₂
MW: 508.56 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00022030
Núm. CAS: 6968-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR