Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate
Proveedor: Apollo Scientific
Sinónimos:
Ethyl 2-[trans-4-(Boc-amino)cyclohexyl]acetate, Ethyl 2-(trans-4-((tert-butoxycarbonyl)amino)cyclohexyl)acetate, 2-[trans-4-(Boc-amino)cyclohexyl]acetic acid Ethyl ester, 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetic acid Ethyl ester
OR470414-1G
OR470414-5G
APOSOR470414-1GEA
136
EUR
APOSOR470414-1G
APOSOR470414-5G
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate
Formula:
C₁₅H₂₇NO₄ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20491405 Núm. CAS: 946598-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...