2-(3-Aminopropoxy)benzonitrile
Proveedor: Apollo Scientific
OR471529-1G
OR471529-5G
APOSOR471529-1GEA
256
EUR
APOSOR471529-1G
APOSOR471529-5G
2-(3-Aminopropoxy)benzonitrile
2-(3-Aminopropoxy)benzonitrile
Formula:
C₁₀H₁₂N₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16495892 Núm. CAS: 444574-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...