N-Boc-D-alanine methyl ester 98%

Proveedor: Apollo Scientific

Sinónimos: Boc-D-Ala-OMe, BOC-D-Alanine methyl ester, (R)-Methyl 2-((tert-butoxycarbonyl)amino)propanoate, BOC-D-Alanine methyl este

OR471627-25G OR471627-100G
APOSOR471627-25GEA 27.7 EUR
APOSOR471627-25G APOSOR471627-100G
N-Boc-D-alanine methyl ester 98%
N-Boc-D-alanine methyl ester
Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191865
Núm. CAS: 91103-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR