N-Benzyloxycarbonylglycine thioamide 97%

Proveedor: Apollo Scientific

OR480692-25G OR480692-5G
APOSOR480692-25GEA 1080 EUR
APOSOR480692-25G APOSOR480692-5G
N-Benzyloxycarbonylglycine thioamide 97%
N-Benzyloxycarbonylglycine thioamide
Formula: C₁₀H₁₂N₂O₂S
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03452777
Núm. CAS: 49548-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR