Propyl-p-toluenesulphonate 99%
Proveedor: Apollo Scientific
|
Atención
|
Sinónimos:
p-Toluenesulphonic acid Propyl Ester
OR55592-25G
OR55592-100G
APOSOR55592-25GEA
109
EUR
APOSOR55592-25G
APOSOR55592-100G
Propyl-p-toluenesulphonate 99%
Propyl-p-toluenesulphonate
Formula:
C₁₀H₁₄O₃S MW: 214,28 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00059244 Núm. CAS: 599-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...