N-Boc-O-methyl-L-threonine 98%
Proveedor: Apollo Scientific
|
Atención
|
Sinónimos:
(2S,3R)-2-(Boc-amino)-3-methoxybutyric acid, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-methoxybutanoic acid, Boc-Thr(Me)-OH
OR61332-1G
OR61332-25G
OR61332-5G
APOSOR61332-1GEA
21.9
EUR
APOSOR61332-1G
APOSOR61332-25G
APOSOR61332-5G
N-Boc-O-methyl-L-threonine 98%
N-Boc-O-methyl-L-threonine
Formula:
C₁₀H₁₉NO₅ MW: 233,3 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00076990 Núm. CAS: 48068-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...