5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine
Proveedor: Apollo Scientific
OR7722-1G
OR7722-5G
APOSOR7722-1GEA
158
EUR
APOSOR7722-1G
APOSOR7722-5G
5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine
5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine
Formula:
C₇H₁₀BrN₃O MW: 232,09 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07781216 Núm. CAS: 57054-81-6 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...