2-Cloro-6-metilquinolina 98%
Proveedor: Apollo Scientific
OR901673-1G
OR901673-5G
APOSOR901673-1GEA
43.5
EUR
APOSOR901673-1G
APOSOR901673-5G
2-Cloro-6-metilquinolina 98%
2-Cloro-6-metilquinolina
Formula:
C₁₀H₈ClN MW: 177,6302 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00462389 Núm. CAS: 4295-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...