6-Chlorohexanoic acid 95%
Proveedor: Apollo Scientific
OR902690-100G
OR902690-25G
APOSOR902690-100GEA
491
EUR
APOSOR902690-100G
APOSOR902690-25G
6-Chlorohexanoic acid 95%
6-Chlorohexanoic acid
Formula:
C₆H₁₁ClO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00061103 Núm. CAS: 4224-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...