5-Methyl-1H-indole-2-carboxylic acid 97%
Proveedor: Apollo Scientific
OR903329-25G
OR903329-5G
APOSOR903329-25GEA
317
EUR
APOSOR903329-25G
APOSOR903329-5G
5-Methyl-1H-indole-2-carboxylic acid 97%
5-Methyl-1H-indole-2-carboxylic acid
Formula:
C10H9NO2 MW: 175,19 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00047166 Núm. CAS: 10241-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...