1,2,3,4-Tetrahydropyrido[2,3-b]pyrazine 95%
Proveedor: Apollo Scientific
OR904722-1G
OR904722-250MG
APOSOR904722-1GEA
87
EUR
APOSOR904722-1G
APOSOR904722-250MG
1,2,3,4-Tetrahydropyrido[2,3-b]pyrazine 95%
1,2,3,4-Tetrahydropyrido[2,3-b]pyrazine
Formula:
C₇H₉N₃ MW: 135,1700 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08062758 Núm. CAS: 35808-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...