2-Amino-5-bromo-3-ethynylpyrazine 95%
Proveedor: Apollo Scientific
OR904952-1G
OR904952-250MG
APOSOR904952-1GEA
353
EUR
APOSOR904952-1G
APOSOR904952-250MG
2-Amino-5-bromo-3-ethynylpyrazine 95%
2-Amino-5-bromo-3-ethynylpyrazine
Formula:
C₆H₄BrN₃ MW: 198,0201 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09999142 Núm. CAS: 1209289-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...