2-Metoxicinamaldehido 95%
Proveedor: Apollo Scientific
OR906162-100G
OR906162-500G
APOSOR906162-100GEA
62.5
EUR
APOSOR906162-100G
APOSOR906162-500G
2-Metoxicinamaldehido 95%
2-Metoxicinamaldehido
Formula:
C₁₀H₁₀O₂ MW: 162,1852 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00007001 Núm. CAS: 1504-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...