2-(4-Methylcyclohexylidene)acetic acid 98%
Proveedor: Apollo Scientific
OR909193-1G
OR909193-25G
OR909193-5G
APOSOR909193-1GEA
125
EUR
APOSOR909193-1G
APOSOR909193-25G
APOSOR909193-5G
2-(4-Methylcyclohexylidene)acetic acid 98%
2-(4-Methylcyclohexylidene)acetic acid
Formula:
C₉H₁₄O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00029539 Núm. CAS: 77842-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...