3,4-Diethoxyaniline 98%
Proveedor: Apollo Scientific
OR911808-1G
OR911808-5G
APOSOR911808-1GEA
0
EUR
APOSOR911808-1G
APOSOR911808-5G
3,4-Diethoxyaniline 98%
3,4-Diethoxyaniline
Formula:
C₁₀H₁₅NO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00026782 Núm. CAS: 39052-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...