N-Cbz-D-phenylalaninol 97%

Proveedor: Apollo Scientific

Sinónimos: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

OR912423-25G
APOSOR912423-25GEA 105 EUR
APOSOR912423-25G
N-Cbz-D-phenylalaninol 97%
N-Cbz-D-phenylalaninol
Formula: C₁₇H₁₉NO₃
MW: 285,3 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191193
Núm. CAS: 58917-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR