4-(Pyridin-4-yl)aniline 95%
Proveedor: Apollo Scientific
OR912614-1G
OR912614-250MG
APOSOR912614-1GEA
0
EUR
APOSOR912614-1G
APOSOR912614-250MG
4-(Pyridin-4-yl)aniline 95%
4-(Pyridin-4-yl)aniline
Formula:
C₁₁H₁₀N₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00970053 Núm. CAS: 13296-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...