4-Butylpiperidine hydrochloride 95%

Proveedor: Apollo Scientific

OR913381-100MG OR913381-1G OR913381-250MG
APOSOR913381-100MGEA 58 EUR
APOSOR913381-100MG APOSOR913381-1G APOSOR913381-250MG
4-Butylpiperidine hydrochloride 95%
4-Butylpiperidine hydrochloride
Formula: C₉H₂₀ClN
MW: 177,72 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09025421
Núm. CAS: 372195-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR