1-Benzylpyrrole 98%
Proveedor: Apollo Scientific
OR917442-25G
OR917442-5G
APOSOR917442-25GEA
287
EUR
APOSOR917442-25G
APOSOR917442-5G
1-Benzylpyrrole 98%
1-Benzylpyrrole
Formula:
C₁₁H₁₁N Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00963301 Núm. CAS: 2051-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...