H-N-Me-Ala-OMe·HCl 95%

Proveedor: Apollo Scientific

Sinónimos: (S)-Methyl 2-(methylamino)propanoate hydrochloride, N-Methyl-l-alanine methyl ester hydrochloride

OR919095-10G OR919095-1G OR919095-5G
APOSOR919095-10GEA 459 EUR
APOSOR919095-10G APOSOR919095-1G APOSOR919095-5G
H-N-Me-Ala-OMe·HCl 95%
H-N-Me-Ala-OMe·HCl
Formula: C₅H₁₁NO₂·HCl
MW: 153,7 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04973252
Núm. CAS: 20045-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR