Ethyl-2-formylpropionate 95%
Proveedor: Apollo Scientific
OR919329-1G
OR919329-250MG
APOSOR919329-1GEA
654
EUR
APOSOR919329-1G
APOSOR919329-250MG
Ethyl-2-formylpropionate 95%
Ethyl-2-formylpropionate
Formula:
C₆H₁₀O₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00044012 Núm. CAS: 27772-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...