Methyl (S)-(-)-2-isocyanato-4-methylvalerate 95%

Proveedor: Apollo Scientific

Sinónimos: (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester

OR919405-1G OR919405-5G
APOSOR919405-1GEA 219 EUR
APOSOR919405-1G APOSOR919405-5G
Methyl (S)-(-)-2-isocyanato-4-methylvalerate 95%
Methyl (S)-(-)-2-isocyanato-4-methylvalerate

  • May form small amount of sediment.

Formula: C₈H₁₃NO₃
MW: 171,20 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191532
Núm. CAS: 39570-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR