trans-3-Hexenedioic acid 95%
Proveedor: Apollo Scientific
OR921177-25G
OR921177-5G
APOSOR921177-25GEA
300
EUR
APOSOR921177-25G
APOSOR921177-5G
trans-3-Hexenedioic acid 95%
trans-3-Hexenedioic acid
Formula:
C₆H₈O₄ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00002785 Núm. CAS: 4436-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...