Methyl-1-naphthoate 95%
Proveedor: Apollo Scientific
Sinónimos:
1-Naphthalenecarboxylic acid methyll ester, 1-Naphthoic acid methyll ester, Methyl 1-Naphthalenecarboxylate, Methyl 1-naphthoate
OR923930-25G
APOSOR923930-25GEA
128
EUR
APOSOR923930-25G
Methyl-1-naphthoate 95%
Methyl-1-naphthoate
Formula:
C₁₂H₁₀O₂ MW: 186,2066 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00094243 Núm. CAS: 2459-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...