Phenyl phosphate 95%

Proveedor: Apollo Scientific

Sinónimos: Phosphoric Acid Phenyl Ester, Dihydrogen Phenyl Phosphate

OR924068-25G OR924068-5G
APOSOR924068-25GEA 261 EUR
APOSOR924068-25G APOSOR924068-5G
Phenyl phosphate 95%
Phenyl phosphate
Formula: C₆H₇O₄P
MW: 174,09 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00058970
Núm. CAS: 701-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR