3-Butoxyphenol 95%
Proveedor: Apollo Scientific
OR924866-25G
OR924866-5G
APOSOR924866-25GEA
818
EUR
APOSOR924866-25G
APOSOR924866-5G
3-Butoxyphenol 95%
3-Butoxyphenol
Formula:
C₁₀H₁₄O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00058976 Núm. CAS: 18979-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...