Methyl angelate 95%
Proveedor: Apollo Scientific
Sinónimos:
Methyl (Z)-2-Methyl-2-butenoate, Methyl 2-Methylisocrotonate, Angelic Acid Methyl Ester, 2-Methylisocrotonic Acid Methyl Ester, (Z)-2-Methyl-2-butenoic Acid Methyl Ester
OR931539-25G
OR931539-5G
APOSOR931539-25GEA
364
EUR
APOSOR931539-25G
APOSOR931539-5G
Methyl angelate 95%
Methyl angelate
Formula:
C₆H₁₀O₂ MW: 114,14 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00063645 Núm. CAS: 5953-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...