(2-Bromoethyl)phosphonic acid 95%
Proveedor: Apollo Scientific
OR932215-1G
APOSOR932215-1GEA
150
EUR
APOSOR932215-1G
(2-Bromoethyl)phosphonic acid 95%
(2-Bromoethyl)phosphonic acid
Formula:
C₂H₆BrO₃P MW: 188,94 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00013957 Núm. CAS: 999-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...