1,1-Dietilurea 97%
Proveedor: Apollo Scientific
OR932849-25G
OR932849-5G
APOSOR932849-25GEA
0
EUR
APOSOR932849-25G
APOSOR932849-5G
1,1-Dietilurea 97%
1,1-Dietilurea
Formula:
C₅H₁₂N₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00007960 Núm. CAS: 634-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...