Alcohol α-ciclopropilbencilico 95%

Proveedor: Apollo Scientific

Sinónimos: alpha-Cyclopropylbenzyl alcohol, Cyclopropyl(phenyl)methanol, α-Cyclopropylbenzyl alcohol, Cyclopropyl phenyl carbinol, Cyclopropylphenylcarbinol

OR936301-1G OR936301-5G
APOSOR936301-1GEA 52.2 EUR
APOSOR936301-1G APOSOR936301-5G
Alcohol α-ciclopropilbencilico 95%
α-Cyclopropylbenzyl alcohol
Formula: C₁₀H₁₂O
MW: 148,2 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001299
Núm. CAS: 1007-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR