Tetramethylhydroquinone 95%

Proveedor: Apollo Scientific

Sinónimos: 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol, 2,3,5,6-Tetramethylbenzene-1,4-diol, Durohydroquinone, Dihydroxydurene

OR936609-1G OR936609-250MG
APOSOR936609-1GEA 149 EUR
APOSOR936609-1G APOSOR936609-250MG
Tetramethylhydroquinone 95%
Tetramethylhydroquinone
Formula: C₁₀H₁₄O₂
MW: 166,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00045781
Núm. CAS: 527-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR