N-Formyl-L-leucine 95%
Proveedor: Apollo Scientific
Sinónimos:
N-Formyl-Leu-OH, For-Leu-OH
OR936620-5G
APOSOR936620-5GEA
72.5
EUR
APOSOR936620-5G
N-Formyl-L-leucine 95%
N-Formyl-L-leucine
Formula:
C₇H₁₃NO₃ MW: 159,19 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00055861 Núm. CAS: 6113-61-7 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...