4-Isopropylcyclohexylamine (cis- and trans- mixture) 95%
Proveedor: Apollo Scientific
Sinónimos:
1-Amino-4-isopropylcyclohexane
OR937443-1G
OR937443-5G
APOSOR937443-1GEA
168
EUR
APOSOR937443-1G
APOSOR937443-5G
4-Isopropylcyclohexylamine (cis- and trans- mixture) 95%
4-Isopropylcyclohexylamine (cis- and trans- mixture)
Formula:
C₉H₁₉N MW: 141.26 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16853065 Núm. CAS: 52430-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...