3-Butilpiridina 98%
Proveedor: Apollo Scientific
OR937482-1G
OR937482-5G
APOSOR937482-1GEA
74.1
EUR
APOSOR937482-1G
APOSOR937482-5G
3-Butilpiridina 98%
3-Butilpiridina
Formula:
C₉H₁₃N MW: 135,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00010709 Núm. CAS: 539-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...