1H-Imidazol-4-ylmethylamine dihydrochloride 97%

Proveedor: Apollo Scientific

OR938259-1G OR938259-250MG
APOSOR938259-1GEA 0 EUR
APOSOR938259-1G APOSOR938259-250MG
1H-Imidazol-4-ylmethylamine dihydrochloride 97%
1H-Imidazol-4-ylmethylamine dihydrochloride
Formula: C₄H₉Cl₂N₃
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09056757
Núm. CAS: 72631-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR