(R)-(+)-Bornylamine 95%

Proveedor: Apollo Scientific

Sinónimos: (R)-bornylamine, Bicyclo[2.2.1]heptan-2-amine,1,7,7-trimethyl-, (1R,2S,4R)-, (4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-amine

OR938471-1G OR938471-250MG
APOSOR938471-1GEA 435 EUR
APOSOR938471-1G APOSOR938471-250MG
(R)-(+)-Bornylamine 95%
(R)-(+)-Bornylamine
Formula: C₁₀H₁₉N
MW: 153,269 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00042691
Núm. CAS: 32511-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR