Boc-D-N-Me-Phg-OH 97%
Proveedor: Apollo Scientific
Sinónimos:
N-Boc-(R)-a-(methylamino)-benzeneacetic acid
OR939352-1G
APOSOR939352-1GEA
40.6
EUR
APOSOR939352-1G
Boc-D-N-Me-Phg-OH 97%
Boc-D-N-Me-Phg-OH
Formula:
C₁₄H₁₉NO₄ MW: 265,3 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07366768 Núm. CAS: 30925-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...