Cyclohexylmethyl acetate 95%

Proveedor: Apollo Scientific

Sinónimos: Acetic Acid Cyclohexylmethyl Ester

OR939832-25G OR939832-5G
APOSOR939832-25GEA 928 EUR
APOSOR939832-25G APOSOR939832-5G
Cyclohexylmethyl acetate 95%
Cyclohexylmethyl acetate
Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 937-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR