3-Azaspiro[5.5]undecane 95%
Proveedor: Apollo Scientific
OR943518-1G
OR943518-250MG
APOSOR943518-1GEA
696
EUR
APOSOR943518-1G
APOSOR943518-250MG
3-Azaspiro[5.5]undecane 95%
3-Azaspiro[5.5]undecane
Formula:
C₁₀H₁₉N Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011572 Núm. CAS: 180-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...